missing translation for 'onlineSavingsMsg'
Learn More

N-alpha-Benzoyl-L-arginine, 99%

Product Code. 11499742
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11499742 10 g 10g
11409752 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11499742 Supplier Thermo Scientific Alfa Aesar Supplier No. A10812.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 154-92-7 | C13H18N4O3 | 278.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 154-92-7
Molecular Formula C13H18N4O3
Molecular Weight (g/mol) 278.31
MDL Number MFCD00001763
InChI Key RSYYQCDERUOEFI-UHFFFAOYNA-N
Synonym bz-arg-oh, n-alpha-benzoyl-l-arginine, n-benzoyl-l-arginine, n2-benzoyl-l-arginine, benzoyl-l-arginine, arginine, n2-benzoyl, nalpha-benzoyl-l-arginine, 2s-5-carbamimidamido-2-phenylformamido pentanoic acid, l-arginine, n2-benzoyl, n∼2∼-phenylcarbonyl-l-arginine
PubChem CID 97369
IUPAC Name (2S)-2-benzamido-5-(diaminomethylideneamino)pentanoic acid
SMILES NC(N)=NCCCC(NC(=O)C1=CC=CC=C1)C(O)=O

Specifications

Melting Point 285°C (decomposition)
Quantity 10 g
Formula Weight 278.31
Percent Purity 99%
Chemical Name or Material N-alpha-Benzoyl-L-arginine

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.