missing translation for 'onlineSavingsMsg'
Learn More

N-alpha-Benzoyl-L-arginine ethyl ester hydrochloride, 98+%

Product Code. 11403707
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11403707

Brand: Thermo Scientific Alfa Aesar A18181.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2645-08-1 | C16H17ClN4O2S | 364.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2645-08-1
MDL Number MFCD00012579

Specifications

Melting Point 127°C to 131°C
Molecular Formula C15H23ClN4O3
Beilstein 3781694
Synonym bz-arg-oet.hcl, n-alpha-benzoyl-l-arginine ethyl ester hydrochloride, ethyl n-benzoyl-l-argininate hydrochloride, baee, nalpha-benzoyl-l-arginine ethyl ester hydrochloride, l-baee, l-arginine, n2-benzoyl-, ethyl ester, monohydrochloride, n-benzoyl-l-arginine ethyl ester hydrochloride, ethyl n2-benzoyl-l-argininate monohydrochloride, ethyl 2s-5-carbamimidamido-2-phenylformamido pentanoate hydrochloride
InChI Key HIXDELXKSSLIKB-UHFFFAOYNA-N
SMILES CCOC(=O)C(CCCN=C(N)N)NC(=O)C1=CC=CC=C1.Cl
IUPAC Name ethyl (2S)-2-benzamido-5-(diaminomethylideneamino)pentanoate;hydrochloride
Molecular Weight (g/mol) 342.82
Quantity 5 g
PubChem CID 2723604
Formula Weight 342.84
Optical Rotation −17° (c=2, Water)
Sensitivity Hygroscopic
Percent Purity ≥98%
Chemical Name or Material N-alpha-Benzoyl-L-arginine ethyl ester hydrochloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.