missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals N-alpha-Benzoyl-L-arginine p-nitroanilide hydrochloride
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
Descripción
N-alpha-Benzoyl-L-arginine p-nitroanilide hydrochloride is used as a chromogenic substrate for trypsin and other proteolytic enzymes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-alpha-Benzoyl-L-arginine p-nitroanilide hydrochloride is used as a chromogenic substrate for trypsin and other proteolytic enzymes.
Solubility
Soluble in DMSO at 50mg/ml
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
N-alpha-Benzoyl-L-arginine p-nitroanilide hydrochloride is used as a chromogenic substrate for trypsin and other proteolytic enzymes.
Solubility
Soluble in DMSO at 50mg/ml
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Especificaciones
Especificaciones
| Melting Point | 223°C |
| Color | White to Yellow |
| Beilstein | 4081878 |
| Solubility Information | Soluble in DMSO at 50mg/ml |
| Quantity | 100 mg |
| Formula Weight | 434.9 |
| Physical Form | Powder |
| Chemical Name or Material | N-alpha-Benzoyl-L-arginine p-nitroanilide hydrochloride |
RUO – Research Use Only
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido