missing translation for 'onlineSavingsMsg'
Learn More

N(alpha)-Boc-D-glutamine, 98+%, Thermo Scientific Chemicals

Product Code. 11378215
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11378215 1 g 1g
11388215 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11378215 Supplier Thermo Scientific Alfa Aesar Supplier No. L08536.03
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 61348-28-5 | Molecular Formula: C10H18N2O5 | Molecular Weight: 246.263 g/mol

N2-Boc-D-glutamine is an N-Boc-protected form of D-Glutamine. D-Glutamine is an unnatural isomer of L-Glutamine that is present in human plasma an is a source of liberated ammonia. D-Glutamine can be synthesized by enzymatic means or can be found in cheeses, wine and vinegars as well.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N2-Boc-D-glutamine is an N-Boc-protected form of D-Glutamine. D-Glutamine is an unnatural isomer of L-Glutamine that is present in human plasma an is a source of liberated ammonia. D-Glutamine can be synthesized by enzymatic means or can be found in cheeses, wine and vinegars as well.

Solubility
Soluble, 1mmol in 2mL DMF. Soluble in chloroform and methanol.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61348-28-5
Molecular Formula C10H18N2O5
Molecular Weight (g/mol) 246.263
MDL Number MFCD00038158
InChI Key VVNYDCGZZSTUBC-ZCFIWIBFSA-N
Synonym boc-d-glutamine, boc-d-gln-oh, boc-d-gln, n alpha-boc-d-glutamine, n-alpha-t-butoxycarbonyl-d-glutamine, n-tert-butoxycarbonyl-d-glutamine, r-5-amino-2-tert-butoxycarbonyl amino-5-oxopentanoic acid, boc-l-gln-oh, n-alpha-t-butyloxycarbonyl-d-glutamine, n-alpha-tert-butyloxycarbonyl-d-glutamine
PubChem CID 2724760
IUPAC Name (2R)-5-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid
SMILES CC(C)(C)OC(=O)NC(CCC(=O)N)C(=O)O

Specifications

Melting Point 117°C to 119°C
Beilstein 1981311
Solubility Information Soluble,1mmol in 2mL DMF. Soluble in chloroform and methanol.
Quantity 1 g
Formula Weight 246.26
Optical Rotation +3.5° (c=2 in Ethanol)
Percent Purity ≥98%
Chemical Name or Material N(alpha)-Boc-D-glutamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.