missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N(alpha)-Boc-D-histidine, 98+%

Product Code. 11352726
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11352726 250 mg 250mg
11342726 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11352726 Supplier Thermo Scientific Chemicals Supplier No. L08810.MD
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 50654-94-9 | C11H17N3O4 | 255.274 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50654-94-9
Molecular Formula C11H17N3O4
Molecular Weight (g/mol) 255.274
MDL Number MFCD00037851
InChI Key AYMLQYFMYHISQO-MRVPVSSYSA-N
Synonym boc-d-his-oh, boc-d-histidine, n-a-boc-d-histidine, d-histidine, n-1,1-dimethylethoxy carbonyl, n-alpha-t-butyloxycarbonyl-d-histidine, r-2-tert-butoxycarbonyl amino-3-1h-imidazol-4-yl propanoic acid, r-2-tert-butoxycarbonyl amino-3-1h-imidazol-4-yl propionic acid, n-alpha-boc-d-histidine, 2r-2-tert-butoxycarbonyl amino-3-1h-imidazol-4-yl propanoic acid, n-boc-d-histidine
PubChem CID 2724761
IUPAC Name (2R)-3-(1H-imidazol-5-yl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CN=CN1)C(=O)O

Specifications

Melting Point ∽197°C (decomposition)
Beilstein 4196575
Quantity 250 mg
Formula Weight 255.27
Optical Rotation −26° (c=1 in Methanol)
Percent Purity ≥98%
Chemical Name or Material N(alpha)-Boc-D-histidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.