missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N(alpha)-Boc-D-tryptophan, 97%

Product Code. 11397286
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397286 1 g 1g
11307296 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11397286 Supplier Thermo Scientific Chemicals Supplier No. L09214.03
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5241-64-5 | C16H20N2O4 | 304.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5241-64-5
Molecular Formula C16H20N2O4
Molecular Weight (g/mol) 304.35
MDL Number MFCD00037944
InChI Key NFVNYBJCJGKVQK-CYBMUJFWSA-N
Synonym boc-d-trp-oh, n-boc-d-tryptophan, n-tert-butoxy carbonyl-d-tryptophan, n-tert-butoxycarbonyl-d-tryptophan, boc-d-tryptophan, boc-d-trp, nalpha-boc-d-tryptophan, boc-d-tryptophane, nalpha-boc-d-tryptophane, tert-butoxycarbonyl
PubChem CID 111050
IUPAC Name (2R)-3-(1H-indol-3-yl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)N[C@H](CC1=CNC2=CC=CC=C12)C(O)=O

Specifications

Melting Point ∽138°C (decomposition)
Beilstein 4237334
Quantity 1 g
Formula Weight 304.35
Optical Rotation +21° (c=1 in Acetic Acid)
Percent Purity 97%
Chemical Name or Material N(alpha)-Boc-D-tryptophan

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.