missing translation for 'onlineSavingsMsg'
Learn More

N(alpha)-Boc-L-2,3-diaminopropionic acid, 97%

Product Code. 15493557
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15493557 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15493557 Supplier Thermo Scientific Alfa Aesar Supplier No. H26919.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 73259-81-1 | C8H16N2O4 | 204.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point ∽210°C (decomposition)
CAS 73259-81-1
Molecular Formula C8H16N2O4
MDL Number MFCD00236843
Synonym boc-dap-oh, 3-amino-boc-l-alanine, boc-l-2,3-diaminopropionic acid, s-3-amino-2-tert-butoxycarbonyl amino propanoic acid, n-boc-beta-amino-ala-oh, boc-dpr-oh, 2s-3-amino-2-tert-butoxy carbonyl amino propanoic acid, 3-amino-n-boc-l-alanine, n-boc-l-2,3-diaminopropanoic acid, nalpha-boc-2,3-diaminopropionic acid
InChI Key KRJLRVZLNABMAT-YFKPBYRVSA-N
SMILES CC(C)(C)OC(=O)N[C@@H](C[NH3+])C([O-])=O
IUPAC Name (2S)-3-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
Molecular Weight (g/mol) 204.23
Quantity 1 g
PubChem CID 2755946
Formula Weight 204.23
Optical Rotation −14° (c=1 in Water)
Percent Purity 97%
Chemical Name or Material N(alpha)-Boc-L-2,3-diaminopropionic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.