missing translation for 'onlineSavingsMsg'
Learn More

N(alpha)-Boc-L-arginine, 98%

Product Code. 15430138
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15430138 1 g 1g
15440138 5 g 5g
15450138 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15430138 Supplier Thermo Scientific Alfa Aesar Supplier No. H53379.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13726-76-6 | C11H22N4O4 | 274.321 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13726-76-6
Molecular Formula C11H22N4O4
Molecular Weight (g/mol) 274.321
MDL Number MFCD00042632
InChI Key HSQIYOPBCOPMSS-ZETCQYMHSA-N
Synonym boc-arg-oh, boc-arginine, s-2-tert-butoxycarbonyl amino-5-guanidinopentanoic acid, boc-l-arg-oh, n2-tert-butoxycarbonyl-l-arginine, n-boc-l-arginine, na-tert-butoxycarbonyl-l-arginine, n-t-boc-l-arginine, n alpha-boc-l-arginine, n∼2∼-tert-butoxycarbonyl-l-arginine
PubChem CID 114667
IUPAC Name (2S)-5-(diaminomethylideneamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
SMILES CC(C)(C)OC(=O)NC(CCCN=C(N)N)C(=O)O

Specifications

Flash Point 113°C (235°F)
Quantity 1 g
Beilstein 2419628
Optical Rotation −6.5° (c=1 in Acetic Acid)
Formula Weight 274.32
Percent Purity 98%
Chemical Name or Material N(alpha)-Boc-L-arginine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.