missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N(alpha)-Boc-L-tryptophan, 98+%

Product Code. 11438656
Change view
Click to view available options
Quantity:
5g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11438656 5g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11438656 Supplier Alfa Aesar™ Supplier No. A16023.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 13139-14-5
Molecular Formula C16H20N2O4
MDL Number MFCD00065595
Beilstein 39677
Synonym boc-trp-oh, n-boc-l-tryptophan, boc-l-tryptophan, n-tert-butoxy carbonyl-l-tryptophan, boc-l-trp-oh, boc-l-tryptophane, s-2-tert-butoxycarbonyl amino-3-1h-indol-3-yl propanoic acid, l-tryptophan, n-1,1-dimethylethoxy carbonyl, boc-tryptophan, n-tert-butoxycarbonyl-l-tryptophan
InChI Key NFVNYBJCJGKVQK-ZDUSSCGKSA-N
SMILES CC(C)(C)OC(=O)NC(CC1=CNC2=CC=CC=C21)C(=O)O
IUPAC Name (2S)-3-(1H-indol-3-yl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
Molecular Weight (g/mol) 304.346
Quantity 5g
PubChem CID 83169
Formula Weight 304.35
Optical Rotation −19.5° (c=1 in DMF)
Percent Purity ≥98%
Chemical Name or Material N(alpha)-Boc-L-tryptophan
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.