missing translation for 'onlineSavingsMsg'
Learn More

N-Amidinothiourea, 98+%, Thermo Scientific™

Product Code. 11497407
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497407 500 g 500g
11477407 25 g 25g
11487407 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11497407 Supplier Thermo Scientific Alfa Aesar Supplier No. A15101.36
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Imino-4-thiobiuret is used in the design and synthesis of guanylthiourea derivatives which are potential inhibitors of plasmodium falciparum dihydrofolate reductase enzyme. 2-Imino-4-thiobiuret is also used as a reagent in recyclization reactions of N-arylmaleimides with polynucleophilic compounds.

Solubility
Soluble in water (70g/L).

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2114-02-5
Molecular Formula C2H6N4S
Molecular Weight (g/mol) 118.16
MDL Number MFCD00014472
InChI Key OKGXJRGLYVRVNE-UHFFFAOYSA-N

Specifications

Melting Point 160°C to 168°C
Odor Odorless
Quantity 500 g
Beilstein 1756620
Synonym amidinothiourea, guanylthiourea, guthimin, guthimine, 2-imino-4-thiobiuret, n-amidinothiourea, thiourea, aminoiminomethyl, thiodicyanodiamidine, diaminomethylidene thiourea, gutimine
Solubility Information Soluble in water (70g/L).
SMILES C(=NC(=S)N)(N)N
IUPAC Name diaminomethylidenethiourea
PubChem CID 2724563
Formula Weight 118.16
Percent Purity ≥98%
Chemical Name or Material N-Amidinothiourea
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.