missing translation for 'onlineSavingsMsg'
Learn More

N-Amyl-N-methylnitrosamine, TRC

Product Code. 30135462
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135462 5 mg 5mg
30134865 50 mg 50mg
30005568 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135462 Supplier Toronto Research Chemicals Supplier No. TRCA6365005MG
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13256-07-0
Molecular Formula C6 H14 N2 O
Molecular Weight (g/mol) 130.19
Synonym N-Nitroso-N-methylpentylamine, 1-Pentanamine, N-methyl-N-nitroso- (9CI, ACI), N-Methyl-N-nitroso-1-pentanamine (ACI), Pentylamine, N-methyl-N-nitroso- (7CI, 8CI), Methylamylnitrosamine, Methylpentylnitrosamine, N-Amyl-N-methylnitrosamine, N-Methyl-N-amylnitrosamine, N-Nitroso-N-methyl-n-pentylamine, N-Nitroso-N-methylamylamine, N-Nitrosomethyl-n-amylamine, N-Nitrosomethylamylamine, Nitrosomethylpentylamine
IUPAC Name N-methyl-N-pentylnitrous amide
SMILES CCCCCN(C)N=O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C6H14N2O/c1-3-4-5-6-8(2)7-9/h3-6H2,1-2H3
Formula Weight 130.1106
Chemical Name or Material N-Amyl-N-methylnitrosamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.