missing translation for 'onlineSavingsMsg'
Learn More

n-Amylamine, 99%

Product Code. 10521842 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
50 mL
250 mL
Packaging:
Glass bottle
Unit Size:
250mL
50mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10521842 5 mL Glass bottle 5mL
10674852 50 mL Glass bottle 50mL
10770965 250 mL Glass bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10521842 Supplier Thermo Scientific Acros Supplier No. 173940050
 Restricted Item
Estimated Shipment: 23-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 110-58-7 | C5H13N | 87.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-58-7
Molecular Formula C5H13N
Molecular Weight (g/mol) 87.15
MDL Number MFCD00008236
InChI Key DPBLXKKOBLCELK-UHFFFAOYSA-N
Synonym amylamine, 1-aminopentane, pentylamine, 1-pentylamine, n-amylamine, 1-pentanamine, n-pentylamine, monoamylamine, norleucamine, amyl amine
PubChem CID 8060
ChEBI CHEBI:74848
IUPAC Name pentan-1-amine
SMILES CCCCCN

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -50.0°C
Color Colorless to Yellow
Density 0.7500g/mL
Boiling Point 104.0°C
Flash Point 4°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)4NH2
Refractive Index 1.4100 to 1.4120
Quantity 5 mL
Beilstein , 175
Merck Index 15, 593
Specific Gravity 0.75
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and ether
Viscosity 1.2 mPa.s (20°C)
Formula Weight 87.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material n-Amylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • n-Amylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.