missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Aspartyl-L-glutamic Acid Dimethyl Ester Trifluoroacetic Acid Salt, TRC
Specifications
Specifications
| Chemical Name or Material | N-Aspartyl-L-glutamic Acid Dimethyl Ester Trifluoroacetic Acid Salt |
| Molecular Formula | C11H18N2O7 . C2HF3O2 |
| Synonym | N-Aspartyl-L-glutamic Acid Dimethyl Ester Trifluoroacetic Acid Salt |
| InChI Formula | InChI=1S/C11H18N2O7.C2HF3O2/c1-19-9(16)4-3-7(11(18)20-2)13-10(17)6(12)5-8(14)15;3-2(4,5)1(6)7/h6-7H,3-5,12H2,1-2H3,(H,13,17)(H,14,15);(H,6,7)/t6-,7-;/m0./s1 |
| SMILES | OC(C[C@H](N)C(N[C@H](C(OC)=O)CCC(OC)=O)=O)=O.OC(C(F)(F)F)=O |
| IUPAC Name | (S)-3-amino-4-(((S)-1,5-dimethoxy-1,5-dioxopentan-2-yl)amino)-4-oxobutanoic acid 2,2,2-trifluoroacetic acid salt |
| Molecular Weight (g/mol) | 404.29 |
| Quantity | 100 mg |
| Formula Weight | 404.1 |
| Shipping Condition | Room Temperature |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?