missing translation for 'onlineSavingsMsg'
Learn More

N-Benzoyl-5'-O-[bis(4-methoxyphenyl)phenylmethyl]-2'-O-(2-methoxyethyl)-adenosine, TRC

Product Code. 30335993
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30335993 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30335993 Supplier Toronto Research Chemicals Supplier No. TRCB207900250MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Benzoyl-5'-O-[bis(4-methoxyphenyl)phenylmethyl]-2'-O-(2-methoxyethyl)-adenosine
CAS 251647-48-0
Molecular Formula C41H41N5O8
Quantity 250 mg
Synonym N-(9-((2R,3R,4R,5R)-5-((bis(4-Methoxyphenyl)(phenyl)methoxy)methyl)-4-hydroxy-3-(2-methoxyethoxy)tetrahydrofuran-2-yl)-9H-purin-6-yl)benzamide, N-Benzoyl-5’-O-DMTR-2’-O-(2-methoxyethyl)-adenosine, 17: PN: US20030211606 PAGE: 19 claimed sequence, 18: PN: US20040005569 PAGE: 22 claimed sequence, 20: PN: US20040006030 PAGE: 23 claimed sequence, 20: PN: US20040014047 PAGE: 21 claimed sequence, 20: PN: US20040014049 PAGE: 21 claimed sequence, 21: PN: US20040005570 PAGE: 20 claimed sequence, 21: PN: US20040014048 PAGE: 21 claimed sequence, 21: PN: US20040014050 PAGE: 20 claimed sequence, 22: PN: US20040005565 PAGE: 17-22 claimed sequence, 22: PN: US20040014051 PAGE: 23 claimed sequence, 23: PN: US20040014699 PAGE: 20 claimed sequence, 24: PN: US20040006029 PAGE: 23 claimed sequence, 24: PN: WO03106645 PAGE: 73 claimed sequence, 95: PN: US20040005707 PAGE: 20 claimed sequence
InChI Formula InChI=1S/C41H41N5O8/c1-49-22-23-52-36-35(47)33(54-40(36)46-26-44-34-37(42-25-43-38(34)46)45-39(48)27-10-6-4-7-11-27)24-53-41(28-12-8-5-9-13-28,29-14-18-31(50-2)19-15-29)30-16-20-32(51-3)21-17-30/h4-21,25-26,33,35-36,40,47H,22-24H2,1-3H3,(H,42,43,45,48)/t33-,35-,36-,40-/m1/s1
SMILES COCCO[C@@H]1[C@H](O)[C@@H](COC(c2ccccc2)(c3ccc(OC)cc3)c4ccc(OC)cc4)O[C@H]1n5cnc6c(NC(=O)c7ccccc7)ncnc56
IUPAC Name N-[9-[(2R,3R,4R,5R)-5-[[bis(4-methoxyphenyl)-phenylmethoxy]methyl]-4-hydroxy-3-(2-methoxyethoxy)oxolan-2-yl]purin-6-yl]benzamide
Molecular Weight (g/mol) 731.79
Formula Weight 731.2955
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.