missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzoylglycine-2,2-d2, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | N-Benzoylglycine-2,2-D2 |
| CAS | 208928-78-3 |
| Molecular Formula | C9 D2 H7 N O3 |
| Quantity | 0.25 g |
| Synonym | Glycine-2,2-d2, N-benzoyl- (9 CI, ACI), N-Benzoylglycine-2,2-d2 (ACI), Hippuric acid-d2 (methylene-d2) |
| InChI Formula | InChI=1 S/C9H9NO3/c11-8(12)6-10-9(13)7-4-2-1-3-5-7/h1-5 H,6H2,(H,10,13)(H,11,12)/i6D2 |
| SMILES | [2 H]C([2 H])(NC(=O)c1ccccc1)C(=O)O |
| IUPAC Name | 2-benzamido-2,2-dideuterioacetic acid |
| Molecular Weight (g/mol) | 181.185 |
| Formula Weight | 181.0708 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction