missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzyl Carvedilol, TRC
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| Chemical Name or Material | (2RS)-1-[benzyl[2-(2-methoxyphenoxy)ethyl]amino]-3-(9H- carbazol-4-yloxy)propan-2-ol |
| CAS | 72955-94-3 |
| Molecular Formula | C31 H32 N2 O4 |
| Quantity | 25 mg |
| Synonym | (2RS)-1-[Benzyl[2-(2-methoxyphenoxy)ethyl]amino]-3-(9H-carbazol-4-yloxy)propan-2-ol, Carvedilol Imp. C (EP) |
| InChI Formula | InChI=1S/C31H32N2O4/c1-35-28-15-7-8-16-29(28)36-19-18-33(20-23-10-3-2-4-11-23)21-24(34)22-37-30-17-9-14-27-31(30)25-12-5-6-13-26(25)32-27/h2-17,24,32,34H,18-22H2,1H3 |
| SMILES | COc1ccccc1OCCN(CC(O)COc2cccc3[nH]c4ccccc4c23)Cc5ccccc5 |
| IUPAC Name | 1-[benzyl-[2-(2-methoxyphenoxy)ethyl]amino]-3-(9H-carbazol-4-yloxy)propan-2-ol |
| Molecular Weight (g/mol) | 496.6 |
| Formula Weight | 496.2362 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction