missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzyl Epinephrine-d3, TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | N-Benzyl Epinephrine-d3 |
| CAS | 1795027-87-0 |
| Molecular Formula | C16H16D3NO3 |
| Quantity | 5 mg |
| Synonym | 4-[1-Hydroxy-2-[methyl(phenylmethyl)amino]ethyl]-1,2-benzenediol-d3, N-Benzyl Epinephrine-d3, rac Adrenaline Impurity D-d3 |
| InChI Formula | InChI=1S/C16H19NO3/c1-17(10-12-5-3-2-4-6-12)11-16(20)13-7-8-14(18)15(19)9-13/h2-9,16,18-20H,10-11H2,1H3/i1D3 |
| SMILES | OC(CN(C([2H])([2H])[2H])CC1=CC=CC=C1)C2=CC=C(O)C(O)=C2 |
| IUPAC Name | 4-[2-[benzyl(trideuteriomethyl)amino]-1-hydroxyethyl]benzene-1,2-diol |
| Molecular Weight (g/mol) | 276.35 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction