missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzyl Hydroxymedetomidine, TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | N-Benzyl Hydroxymedetomidine |
| CAS | 2250243-44-6 |
| Molecular Formula | C20H22N2O |
| Quantity | 5 mg |
| Synonym | 1-(1-Benzyl-1H-imidazol-5-yl)-1-(2,3-dimethylphenyl)ethanol, Medetomidine Impurity 21, alpha-(2,3-Dimethylphenyl)-alpha-methyl-1-(phenylmethyl)-1H-imidazole-5-methanol |
| InChI Formula | InChI=1S/C20H22N2O/c1-15-8-7-11-18(16(15)2)20(3,23)19-12-21-14-22(19)13-17-9-5-4-6-10-17/h4-12,14,23H,13H2,1-3H3 |
| SMILES | Cc1cccc(c1C)C(C)(O)c2cncn2Cc3ccccc3 |
| IUPAC Name | 1-(3-benzylimidazol-4-yl)-1-(2,3-dimethylphenyl)ethanol |
| Molecular Weight (g/mol) | 306.4 |
| Formula Weight | 306.1732 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction