missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl Hydroxymedetomidine, TRC

Product Code. 30331044
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 30331044

Brand: Toronto Research Chemicals TRCB28002550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Benzyl Hydroxymedetomidine
CAS 2250243-44-6
Molecular Formula C20H22N2O
Quantity 50 mg
Synonym 1-(1-Benzyl-1H-imidazol-5-yl)-1-(2,3-dimethylphenyl)ethanol, Medetomidine Impurity 21, alpha-(2,3-Dimethylphenyl)-alpha-methyl-1-(phenylmethyl)-1H-imidazole-5-methanol
InChI Formula InChI=1S/C20H22N2O/c1-15-8-7-11-18(16(15)2)20(3,23)19-12-21-14-22(19)13-17-9-5-4-6-10-17/h4-12,14,23H,13H2,1-3H3
SMILES Cc1cccc(c1C)C(C)(O)c2cncn2Cc3ccccc3
IUPAC Name 1-(3-benzylimidazol-4-yl)-1-(2,3-dimethylphenyl)ethanol
Molecular Weight (g/mol) 306.4
Formula Weight 306.1732
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.