missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl-N-ethylaniline, 98%, Thermo Scientific™

Product Code. 11448717
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11448717

Brand: Thermo Scientific Alfa Aesar A19882.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-59-1
Molecular Formula C15H17N
Molecular Weight (g/mol) 211.31
MDL Number MFCD00009037
InChI Key HSZCJVZRHXPCIA-UHFFFAOYSA-N
Synonym ethylbenzylaniline, benzenemethanamine, n-ethyl-n-phenyl, n-ethyl-n-phenylbenzylamine, benzylethylphenylamine, phenylethylbenzylamine, n-ethyl-n-benzylaniline, benzylamine, n-ethyl-n-phenyl, n-ethyl-n-phenyl benzylamine, n-ethyl-n-phenylbenzenemethanamine, amine, benzyl ethyl phenyl
PubChem CID 7098
IUPAC Name N-benzyl-N-ethylaniline
SMILES CCN(CC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Density 1.03
Boiling Point 163°C to 164°C (6 mmHg)
Flash Point >157°C (315°F)
Refractive Index 1.595
Quantity 10 g
UN Number UN2274
Beilstein 2099852
Merck Index 14,3769
Formula Weight 211.31
Percent Purity 98%
Chemical Name or Material N-Benzyl-N-ethylaniline
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.