missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl (-)-Nebivolol, TRC

Product Code. 30011044
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011044 1 mg 1mg
30012431 5 mg 5mg
30009028 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011044 Supplier Toronto Research Chemicals Supplier No. TRCB2857331MG
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1199945-26-0
Molecular Formula C29H31F2NO4
Molecular Weight (g/mol) 495.56
Synonym (αS,α'S,2R,2'S)-α,α'-[[(Phenylmethyl)imino]bis(methylene)]bis[6-fluoro-3,4-dihydro-2H-1-benzopyran-2-methanol;
IUPAC Name (1S)-2-[benzyl-[(2S)-2-[(2S)-6-fluoro-3,4-dihydro-2H-chromen-2-yl]-2-hydroxyethyl]amino]-1-[(2R)-6-fluoro-3,4-dihydro-2H-chromen-2-yl]ethanol
SMILES C1CC2=C(C=CC(=C2)F)O[C@H]1[C@H](CN(CC3=CC=CC=C3)C[C@@H]([C@@H]4CCC5=C(O4)C=CC(=C5)F)O)O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C29H31F2NO4/c30-22-8-12-26-20(14-22)6-10-28(35-26)24(33)17-32(16-19-4-2-1-3-5-19)18-25(34)29-11-7-21-15-23(31)9-13-27(21)36-29/h1-5,8-9,12-15,24-25,28-29,33-34H,6-7,10-11,16-18H2/t24-,25-,28-,29+/m0/s1
Chemical Name or Material N-Benzyl (-)-Nebivolol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.