missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl Nortropinone, TRC

Product Code. 30328878
Change view
Click to view available options
Quantity:
2 g
Unit Size:
2g
Product Code. Quantity unitSize
30328878 2 g 2g
1 options
This item is not returnable. View return policy

Product Code. 30328878

Brand: Toronto Research Chemicals TRCB2858102G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Benzylnortropinone
CAS 28957-72-4
Molecular Formula C14 H17 N O
Quantity 2 g
InChI Formula InChI=1S/C14H17NO/c16-14-8-12-6-7-13(9-14)15(12)10-11-4-2-1-3-5-11/h1-5,12-13H,6-10H2/t12-,13+
SMILES O=C1C[C@H]2CC[C@@H](C1)N2Cc3ccccc3
IUPAC Name (1R,5S)-8-benzyl-8-azabicyclo[3.2.1]octan-3-one
Molecular Weight (g/mol) 215.29
Formula Weight 215.131
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.