missing translation for 'onlineSavingsMsg'
Learn More

N-Benzylglycine ethyl ester, 97%

Product Code. 11308286
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11308286

Brand: Thermo Scientific Alfa Aesar L15496.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6436-90-4 | C11H15NO2 | 193.25 g/mol

N-Benzylglycine ethyl ester is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Benzylglycine ethyl ester is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6436-90-4
Molecular Formula C11H15NO2
Molecular Weight (g/mol) 193.25
MDL Number MFCD00009174
InChI Key ULOLIZHBYWAICY-UHFFFAOYSA-N
Synonym n-benzylglycine ethyl ester, ethyl 2-benzylamino acetate, ethyl n-benzylglycinate, n-bzl-gly-oet, bzl-gly-oet, glycine, n-phenylmethyl-, ethyl ester, n-benzyl glycine ethyl ester, ethyl n-phenylmethyl glycinate, ethyl benzylamino acetate, n-benzyl-glycine ethyl ester
PubChem CID 80908
IUPAC Name ethyl 2-(benzylamino)acetate
SMILES CCOC(=O)CNCC1=CC=CC=C1

Specifications

Boiling Point 102°C to 104°C (1mmHg)
Refractive Index 1.5053
Beilstein 2104816
Solubility Information Not miscible or difficult to mix in water.
Density 1.048
Quantity 25 g
Flash Point 112°C (233°F)
Formula Weight 193.25
Percent Purity 97%
Chemical Name or Material N-Benzylglycine ethyl ester

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.