missing translation for 'onlineSavingsMsg'
Learn More

N-Benzylidenemethylamine, 99%

Product Code. 10735171 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10735171

Brand: Thermo Scientific Acros 106010250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 622-29-7 | C8H9N | 119.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-29-7
MDL Number MFCD00008294
InChI Key HXTGGPKOEKKUQO-UHFFFAOYSA-N
Synonym n-benzylidenemethylamine, n-benzylidenemethanamine, benzylidenemethylamine, n-methylbenzaldimine, methanamine, n-phenylmethylene, benzylidenemethanamine, n-methylbenzylideneamine, methylamine, n-benzylidene, n-phenylmethylene methanamine, benzylidene-methyl-amine
PubChem CID 73954
IUPAC Name N-methyl-1-phenylmethanimine
SMILES CN=CC1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color White
Density 0.9600g/mL
Boiling Point 75°C to 80°C (18.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C8H9N
Linear Formula C6H5CH=NCH3
Refractive Index 1.5510 to 1.553
Quantity 25 g
Beilstein 07, 213
Specific Gravity 0.96
Molecular Weight (g/mol) 119.17
Formula Weight 119.17
Percent Purity 99%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material N-Benzylidenemethylamine, 99%
Show More Show Less
compliance-icons
Product Identifier
  • N-Benzylidenemethylamine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.