missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-1,3-diaminopropane hydrochloride, 97%, Thermo Scientific™

Product Code. 15448126
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15448126 5 g 5g
15438126 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15448126 Supplier Thermo Scientific Alfa Aesar Supplier No. H63103.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17400-34-9
Molecular Formula C11H17ClN2O2
Molecular Weight (g/mol) 244.72
MDL Number MFCD00270153
InChI Key XKMBTMXQMDLSRB-UHFFFAOYSA-N
Synonym n-cbz-1,3-diaminopropane hydrochloride, benzyl 3-aminopropyl carbamate hydrochloride, benzyl n-3-aminopropyl carbamate hydrochloride, n-cbz-1,3-diaminopropane-hcl, n-z-1,3-propanediamine hydrochloride, n-carbobenzoxy-1,3-diaminopropane hydrochloride, n-carbobenzyloxy-1,3-diaminopropane hydrochloride, n-3-aminopropyl carbamic acid benzyl ester hydrochloride, n-1-z-1,3-diaminopropane hcl, n-z-1,3-diaminopropane hydrochloride
PubChem CID 13196227
IUPAC Name benzyl N-(3-aminopropyl)carbamate;hydrochloride
SMILES [H+].[Cl-].NCCCNC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 185°C to 189°C
Color White
Quantity 5 g
Beilstein 4725495
Sensitivity Hygroscopic
Formula Weight 244.72
Percent Purity 97%
Physical Form Crystalline Solid
Chemical Name or Material N-Benzyloxycarbonyl-1,3-diaminopropane hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.