Learn More
N-Benzyloxycarbonyl-1,3-propanediamine hydrochloride, 98%
Shop All Thermo Scientific Chemicals ProductsDescription
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 97.5 |
| CAS Max % | 100.0 |
| Melting Point | 185°C to 189°C |
| Color | Yellow |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98% |
| Packaging | Glass bottle |
| Quantity | 1 g |
| IUPAC Name | hydrogen benzyl N-(3-aminopropyl)carbamate chloride |
| Formula Weight | 244.72 |
| Show More |
Safety and Handling
- N-Benzyloxycarbonyl-1,3-propanediamine hydrochloride
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.