missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-D-phenylalanine, 98+%, Thermo Scientific™

Product Code. 11377226
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11377226 5 g 5g
11367226 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11377226 Supplier Thermo Scientific Alfa Aesar Supplier No. L09102.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
N-Benzyloxycarbonyl-D-phenylalanine is used for affinity chromatography of proteinases such as the extracellular fibrinogenolytic enzyme of Aspergillus fumigatus. It serves as a primary precursor for the biosynthesis of catecholamines in the body. Also used to antagonize stress-induced analgesia in humans, and also acts as an anti-enkephalinase agent.

Solubility
Soluble in methanol. Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2448-45-5
Molecular Formula C17H17NO4
Molecular Weight (g/mol) 299.326
MDL Number MFCD00063151
InChI Key RRONHWAVOYADJL-OAHLLOKOSA-N
Synonym z-d-phe-oh, n-cbz-d-phenylalanine, z-d-phenylalanine, cbz-d-phe-oh, n-carbobenzyloxy-d-phenylalanine, chembl63865, benzyloxycarbonyl-d-phenylalanine, n-benzyloxycarbonyl-d-phenylalanine, 2r-3-phenyl-2-phenylmethoxycarbonylamino propanoic acid, d-phenylalanine, n-phenylmethoxy carbonyl
PubChem CID 675678
IUPAC Name (2R)-3-phenyl-2-(phenylmethoxycarbonylamino)propanoic acid
SMILES C1=CC=C(C=C1)CC(C(=O)O)NC(=O)OCC2=CC=CC=C2

Specifications

Melting Point 85°C to 87°C
Quantity 5 g
Beilstein 2817463
Solubility Information Soluble in methanol. Slightly soluble in water.
Optical Rotation −5° (c=5 in Acetic Acid)
Formula Weight 299.33
Percent Purity ≥98%
Chemical Name or Material N-Benzyloxycarbonyl-D-phenylalanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.