missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-D-threonine, 98+%, Thermo Scientific™

Product Code. 11322586
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11322586 250 mg 250mg
11312586 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11322586 Supplier Thermo Scientific Alfa Aesar Supplier No. L08561.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80384-27-6
Molecular Formula C12H15NO5
Molecular Weight (g/mol) 253.25
MDL Number MFCD00037808
InChI Key IPJUIRDNBFZGQN-WCBMZHEXSA-N
Synonym z-d-thr-oh, z-d-threonine, n-carbobenzoxy-d-threonine, 2r,3s-2-benzyloxy carbonyl amino-3-hydroxybutanoic acid, n-benzyloxycarbonyl-d-threonine, n-cbz-d-threonine, d-threonine, n-phenylmethoxy carbonyl, 2r,3s-3-hydroxy-2-phenylmethoxycarbonylamino butanoic acid, cbz-d-thr-oh
PubChem CID 853484
IUPAC Name (2R,3S)-3-hydroxy-2-(phenylmethoxycarbonylamino)butanoic acid
SMILES C[C@H](O)[C@@H](NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 100°C to 105°C
Quantity 250 mg
Beilstein 4695777
Optical Rotation +5.5° (c=2 in Acetic Acid)
Formula Weight 253.26
Percent Purity ≥98%
Chemical Name or Material N-Benzyloxycarbonyl-D-threonine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.