missing translation for 'onlineSavingsMsg'
Learn More

N-(Benzyloxycarbonyl)ethylenediamine Hydrochloride, TRC

Product Code. 30336668
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30336668 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30336668 Supplier Toronto Research Chemicals Supplier No. TRCB28728025G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(Benzyloxycarbonyl)ethylenediamine Hydrochloride
CAS 18807-71-1
Molecular Formula C10H14N2O2 • HCl
Quantity 25 g
Synonym N-[(Benzyloxy)carbonyl]-1,2-diaminoethane Hydrochloride, N-(Benzyloxycarbonyl)ethane-1,2-diamine Hydrochloride, N-(2-Aminoethyl)carbamic Acid Benzyl Ester Hydrochloride, Benzyl (2-Aminoethyl)carbamate Hydrochloride, 2-[[(Benzyloxy)carbonyl]amino]ethanamine Hydrochloride, 2-(Benzyloxycarbonylamino)ethylamine Hydrochloride, 1-(Benzyloxycarbonylamino)-2-aminoethane Hydrochloride, (2-Aminoethyl)carbamic Acid Benzyl Ester Hydrochloride, (2-Aminoethyl)carbamic Acid Phenylmethyl Ester Monohydrochloride, (2-Aminoethyl)carbamic Acid Benzyl Ester Monohydrochloride
InChI Formula InChI=1S/C10H14N2O2.ClH/c11-6-7-12-10(13)14-8-9-4-2-1-3-5-9;/h1-5H,6-8,11H2,(H,12,13);1H
SMILES NCCNC(OCC1=CC=CC=C1)=O.Cl
IUPAC Name benzyl N-(2-aminoethyl)carbamate;hydrochloride
Molecular Weight (g/mol) 194.233646
Formula Weight 230.0822
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.