missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-L-glutamic acid 5-tert-butyl ester, 97%, Thermo Scientific Chemicals

Product Code. 11495127
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11495127 5 g 5g
11485127 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 11495127

Brand: Thermo Scientific Alfa Aesar B22557.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3886-08-6
Molecular Formula C17H22NO6
Molecular Weight (g/mol) 336.37
MDL Number MFCD00038274
InChI Key GLMODRZPPBZPPB-ZDUSSCGKSA-M
Synonym z-glu otbu-oh, cbz-glu obut-oh, cbz-l-glu otbu-oh, s-2-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid, n-cbz-l-glutamic acid 5-tert-butyl ester, n-benzyloxycarbonyl-l-glutamic acid 5-tert-butyl ester, 2s-2-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid, 2s-4-tert-butyl oxycarbonyl-2-phenylmethoxy carbonylamino butanoic acid, 5-tert-butyl n-carbobenzoxy-l-glutamate, pubchem6261
PubChem CID 107472
IUPAC Name (2S)-5-[(2-methylpropan-2-yl)oxy]-5-oxo-2-(phenylmethoxycarbonylamino)pentanoic acid
SMILES CC(C)(C)OC(=O)CCC(C(=O)O)NC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 85°C to 88°C
Quantity 5 g
Optical Rotation −13° (c=2 in Methanol)
Formula Weight 337.37
Percent Purity 97%
Chemical Name or Material N-Benzyloxycarbonyl-L-glutamic acid 5-tert-butyl ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.