missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-L-serine, 99%

Product Code. 11419356
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. Quantity unitSize
11419356 5 g 5g
11429356 25 g 25g
11439356 100 g 100g
3 options
This item is not returnable. View return policy

Product Code. 11419356

Brand: Thermo Scientific Alfa Aesar A19050.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1145-80-8 | C11H13NO5 | 239.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1145-80-8
Molecular Formula C11H13NO5
Molecular Weight (g/mol) 239.23
MDL Number MFCD00002662
InChI Key GNIDSOFZAKMQAO-UHFFFAOYNA-N
Synonym z-ser-oh, n-cbz-l-serine, benzyloxycarbonylserine, z-l-serine, carbobenzyloxy-l-serine, n-carbobenzyloxy-l-serine, cbz-l-ser-oh, n-carbobenzoxy-l-serine, n-benzyloxycarbonyl-l-serine, cbz-l-serine
PubChem CID 100310
IUPAC Name (2S)-3-hydroxy-2-(phenylmethoxycarbonylamino)propanoic acid
SMILES OCC(NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 117°C to 120°C
Quantity 5 g
Beilstein 2058314
Optical Rotation +6° (c=7 in Acetic Acid)
Formula Weight 239.23
Percent Purity 99%
Chemical Name or Material N-Benzyloxycarbonyl-L-serine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.