missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-L-serine benzyl ester, 99%

Product Code. 11485257
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11485257

Brand: Thermo Scientific Alfa Aesar B22647.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 21209-51-8 | C18H19NO5 | 329.352 g/mol

N-Benzyloxycarbonyl-L-serine benzyl ester is used as a building block in peptide synthesis and as a biochemical reagent. It is also used to synthesize the corresponding N-benzyloxycarbonyl-O-tert-butyl-(lor dl)-serine benzyl ester. Further, it serves as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Benzyloxycarbonyl-L-serine benzyl ester is used as a building block in peptide synthesis and as a biochemical reagent. It is also used to synthesize the corresponding N-benzyloxycarbonyl-O-tert-butyl-(lor dl)-serine benzyl ester. Further, it serves as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21209-51-8
Molecular Formula C18H19NO5
Molecular Weight (g/mol) 329.352
MDL Number MFCD00037824
InChI Key MHHDPGHZHFJLBZ-INIZCTEOSA-N
Synonym z-ser-obzl, cbz-ser-obzl, n-cbz-l-serine benzyl ester, n-z-l-serine benzyl ester, s-benzyl 2-benzyloxy carbonyl amino-3-hydroxypropanoate, n-carbobenzoxy-l-serine benzyl ester, benzyl n-benzyloxy carbonyl-l-serinate, l-serine,n-phenylmethoxy carbonyl-, phenylmethyl ester, serine, n-phenylmethoxy carbonyl-, phenylmethyl ester, z-l-serine benzyl ester
PubChem CID 6992970
IUPAC Name benzyl (2S)-3-hydroxy-2-(phenylmethoxycarbonylamino)propanoate
SMILES C1=CC=C(C=C1)COC(=O)C(CO)NC(=O)OCC2=CC=CC=C2

Specifications

Melting Point 82°C to 84°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 329.32
Percent Purity 99%
Chemical Name or Material N-Benzyloxycarbonyl-L-serine benzyl ester

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.