missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-L-tyrosine methyl ester, 98%

Product Code. 15479618
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15479618 5 g 5g
15489618 25 g 25g
15418136 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15479618 Supplier Thermo Scientific Alfa Aesar Supplier No. H63113.06
 Restricted Item
Estimated Shipment: 18-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13512-31-7 | C18H19NO5 | 329.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13512-31-7
Molecular Formula C18H19NO5
Molecular Weight (g/mol) 329.35
MDL Number MFCD00038784
InChI Key SLLMDHBKALJDBW-XISACWJONA-N
Synonym z-tyr-ome, z-l-tyrosine methyl ester, n-cbz-l-tyrosine methyl ester, s-methyl 2-benzyloxy carbonyl amino-3-4-hydroxyphenyl propanoate, cbz-tyr-ome, z-l-tyr-ome, z-tyr-och3, cbz-tyrosine methyl ester, n-cbz-l-tyrosinemethylester, n-benzyloxycarbonyltyrosine methyl ester
PubChem CID 7010613
SMILES COC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 93°C to 97°C (decomposition)
IUPAC Name methyl 2-{[(benzyloxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoate
Quantity 5 g
Formula Weight 329.35
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Benzyloxycarbonyl-L-tyrosine methyl ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.