missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyltropinone, 97%, Thermo Scientific™

Product Code. 15352022 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15352022 5 g 5g
15362022 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15352022 Supplier Thermo Scientific Acros Supplier No. 269870050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 28957-72-4
Molecular Formula C14H17NO
Molecular Weight (g/mol) 215.30
MDL Number MFCD00151640
InChI Key RSUHKGOVXMXCND-UHFFFAOYNA-N
Synonym 8-benzyl-8-azabicyclo 3.2.1 octan-3-one, n-benzylnortropinone, n-benzyltropinone, 8-benzyl-8-aza-bicyclo 3.2.1 octan-3-one, 8-phenylmethyl-8-azabicyclo 3.2.1 octan-3-one, 8-azabicyclo 3.2.1 octan-3-one, 8-phenylmethyl, n-benzyl tropinone, 8-benzyl-8-azabicyclo 3.2.1 octan-3-on, n-benzyl-8-azabicyclo 3.2.1 octan-3-one
PubChem CID 119846
IUPAC Name 8-benzyl-8-azabicyclo[3.2.1]octan-3-one
SMILES O=C1CC2CCC(C1)N2CC1=CC=CC=C1

Specifications

Melting Point 28.5°C to 30.5°C
Assay Percent Range 97%
Refractive Index 1.556
Quantity 5 g
Formula Weight 215.29
Percent Purity 97%
Chemical Name or Material N-Benzyltropinone
compliance-icons
Product Identifier
  • N-Benzyltropinone, 97%
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.