missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-1,2,5,6-tetrahydropyridine-4-boronic acid pinacol ester, 95%

Product Code. 15492699
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15492699 5 g 5g
15482699 1 g 1g
15402709 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15492699 Supplier Thermo Scientific Alfa Aesar Supplier No. H66193.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 286961-14-6 | C16H28BNO4 | 309.21 g/mol

Reagent used for suzuki-Miyaura cross-coupling using palladium phosphine catalyst, palladium-catalyzed ligand-controlled regioselective Suzuki coupling, wrenchnolol derivative optimized for gene activation in cells. Also used in Preparation of several enzymatic inhibitors and receptor ligands, orally active anaplastic lymphoma kinase inhibitors and oxazolecarboxamides as diacylglycerol acyltransferase-1 inhibitors for treatment of obesity and diabetes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent used for suzuki-Miyaura cross-coupling using palladium phosphine catalyst, palladium-catalyzed ligand-controlled regioselective Suzuki coupling, wrenchnolol derivative optimized for gene activation in cells. Also used in Preparation of several enzymatic inhibitors and receptor ligands, orally active anaplastic lymphoma kinase inhibitors and oxazolecarboxamides as diacylglycerol acyltransferase-1 inhibitors for treatment of obesity and diabetes.

Solubility
Soluble in most organic solvents. Insoluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 286961-14-6
Molecular Formula C16H28BNO4
Molecular Weight (g/mol) 309.21
MDL Number MFCD03840345
InChI Key VVDCRJGWILREQH-UHFFFAOYSA-N
Synonym n-boc-1,2,5,6-tetrahydropyridine-4-boronic acid pinacol ester, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-5,6-dihydropyridine-1 2h-carboxylate, 1-n-boc-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,6-dihydro-2h-pyridine, n-boc-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydro-2h-pyridine-1-carboxylate, tert-butyl 5,6-dihydro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-1 2h-carboxylate, 3,6-dihydro-2h-pyridine-1-n-boc-4-boronic acid pinacol ester, n-tert-butoxycarbonyl-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester, 1-boc-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,6-dihydro-2h-pyridine, tert-butyl 4-tetramethyl-1,3,2-dioxaborolan-2-yl-1,2,3,6-tetrahydropyridine-1-carboxylate
PubChem CID 4642098
IUPAC Name tert-butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,6-dihydro-2H-pyridine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(=CC1)B1OC(C)(C)C(C)(C)O1

Specifications

Quantity 5 g
Solubility Information Soluble in most organic solvents. Insoluble in water.
Formula Weight 327.23
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Boc-1,2,5,6-tetrahydropyridine-4-boronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.