missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-1,6-diaminohexane, 98%

Product Code. 11399955
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
11399955 5 g 5g
11389955 1 g 1g
11309965 25 g 25g
3 options
This item is not returnable. View return policy

Product Code. 11399955

Brand: Thermo Scientific Alfa Aesar L19948.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 51857-17-1 | C11H24N2O2 | 216.325 g/mol

N-Boc-1,6-diaminohexane is used to prepare 1,3-Bis[6-(Boc-amino)hexyl]urea by reacting with carbonyl dichloride in the presence of triethylamine. Further, it is used as a reagent for the introduction of a C6-spacer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-1,6-diaminohexane is used to prepare 1,3-Bis[6-(Boc-amino)hexyl]urea by reacting with carbonyl dichloride in the presence of triethylamine. Further, it is used as a reagent for the introduction of a C6-spacer.

Solubility
Soluble in dichloromethane and ethyl acetate.

Notes
Air sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51857-17-1
Molecular Formula C11H24N2O2
Molecular Weight (g/mol) 216.325
MDL Number MFCD00671489
InChI Key RVZPDKXEHIRFPM-UHFFFAOYSA-N
Synonym n-boc-1,6-diaminohexane, n-boc-1,6-hexanediamine, tert-butyl n-6-aminohexyl carbamate, tert-butyl 6-aminohexyl carbamate, n-tert-butoxycarbonyl-1,6-diaminohexane, n-tert-butoxycarbonyl-1,6-hexanediamine, tert-butyl 6-aminohexylcarbamate, boc-1,6-diaminohexane hydrochloride, 6-tert-butoxycarbonylamino hexylamine, n-6-aminohexyl carbamic acid tert-butyl ester
PubChem CID 2733170
IUPAC Name tert-butyl N-(6-aminohexyl)carbamate
SMILES CC(C)(C)OC(=O)NCCCCCCN

Specifications

Boiling Point 106°C to 110°C (0.3mmHg)
Refractive Index 1.461
UN Number UN2735
Beilstein 2089264
Solubility Information Soluble in dichloromethane and ethyl acetate.
Density 0.965
Quantity 5 g
Flash Point >110°C (230°F)
Formula Weight 216.33
Sensitivity Air Sensitive
Percent Purity 98%
Chemical Name or Material N-Boc-1,6-diaminohexane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.