missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-1,6-diaminohexane hydrochloride, 98%

Product Code. 11368836
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11368836

Brand: Thermo Scientific Alfa Aesar L19487.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 65915-94-8 | C11H25ClN2O2 | 252.78 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65915-94-8
Molecular Formula C11H25ClN2O2
Molecular Weight (g/mol) 252.78
MDL Number MFCD00039072
InChI Key JSBWQIZQJOQPFN-UHFFFAOYSA-N
Synonym n-boc-1,6-hexanediamine hydrochloride, tert-butyl 6-aminohexyl carbamate hydrochloride, n-boc-1,6-diamino-hexane hydrochloride, tert-butyl n-6-aminohexyl carbamate hydrochloride, n-6-aminohexyl tert-butoxy carboxamide, chloride, tert-butyl 6-aminohexyl carbamate monohydrochloride, tert-butyl 6-aminohexyl carbamate hcl, acmc-1b5cm, n-1-boc-1,6-diaminohexane . hcl, n-boc-1,6-hexanediamine hcl
PubChem CID 3017638
IUPAC Name tert-butyl N-(6-aminohexyl)carbamate;hydrochloride
SMILES [H+].[Cl-].CC(C)(C)OC(=O)NCCCCCCN

Specifications

Melting Point ∽164°C (decomposition)
Beilstein 3631740
Quantity 5 g
Formula Weight 252.79
Sensitivity Hygroscopic
Percent Purity 98%
Chemical Name or Material N-Boc-1,6-diaminohexane hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.