missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-2-methyl-D-serine, 97%, Thermo Scientific Chemicals

Product Code. 15499527
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15499527 1 g 1g
15409537 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15499527 Supplier Thermo Scientific Alfa Aesar Supplier No. H52787.03
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84311-18-2
Molecular Formula C9H17NO5
Molecular Weight (g/mol) 219.237
MDL Number MFCD02682596
InChI Key FWRXDSRYWWYTPD-SECBINFHSA-N
Synonym r-2-tert-butoxycarbonyl amino-3-hydroxy-2-methylpropanoic acid, n-boc-alpha-methyl-d-serine, boc-alpha-methyl-d-ser, boc-me-d-ser-oh, 2r-2-tert-butoxycarbonyl amino-3-hydroxy-2-methylpropanoic acid, boc-d-alpha-methylserine, boc-a-methyl-d-ser, n-boc-a-methyl-d-serine, n-boc-2-methyl-d-serine, n-tert-butoxycarbonyl-2-methyl-d-serine
PubChem CID 24749404
IUPAC Name (2R)-3-hydroxy-2-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(C)(CO)C(=O)O

Specifications

Quantity 1 g
Formula Weight 219.24
Percent Purity 97%
Chemical Name or Material N-Boc-2-methyl-D-serine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.