missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-2-methyl-D-serine methyl ester, 97%, Thermo Scientific Chemicals

Product Code. 15489147
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15489147 1 g 1g
15499147 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15489147 Supplier Thermo Scientific Alfa Aesar Supplier No. H52421.03
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 188476-33-7
Molecular Formula C10H19NO5
Molecular Weight (g/mol) 233.264
MDL Number MFCD06797553
InChI Key OUUNEDPIBZNRMT-SNVBAGLBSA-N
Synonym n-boc-alpha-methyl-d-serine methyl ester, n-boc-2-methyl-d-serine methyl ester, n-tert-butoxycarbonyl-2-methyl-d-serine methyl ester, r-methyl 2-tert-butoxycarbonyl amino-3-hydroxy-2-methylpropanoate, methyl 2r-2-tert-butoxycarbonyl amino-3-hydroxy-2-methylpropanoate
PubChem CID 10752097
IUPAC Name methyl (2R)-3-hydroxy-2-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
SMILES CC(C)(C)OC(=O)NC(C)(CO)C(=O)OC

Specifications

Quantity 1 g
Formula Weight 233.26
Percent Purity 97%
Chemical Name or Material N-Boc-2-methyl-D-serine methyl ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.