missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-2-propargyl-D-glycine, 95%, Thermo Scientific™

Product Code. 15499078
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15499078 1 g 1g
15409088 250 mg 250mg
2 options
This item is not returnable. View return policy

Product Code. 15499078

Brand: Thermo Scientific Alfa Aesar H62572.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63039-46-3
Molecular Formula C10H14NO4
Molecular Weight (g/mol) 212.23
MDL Number MFCD01320886
InChI Key AMKHAJIFPHJYMH-SSDOTTSWSA-M
Synonym boc-d-propargylglycine, r-2-tert-butoxycarbonyl amino pent-4-ynoic acid, n-boc-2-propargyl-d-glycine, boc-d-pra-oh, s-n-boc-propargylglycine, 2r-2-tert-butoxycarbonyl amino pent-4-ynoic acid, boc-d-gly propargyl-oh, boc-r-2-propargylglycine, n-tert-butoxycarbonyl-3-ethynyl-d-alanine, 2-r-tert-butoxycarbonylaminopent-4-ynoic acid
PubChem CID 7009126
SMILES CC(C)(C)OC(=O)N[C@H](CC#C)C([O-])=O

Specifications

Quantity 1 g
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pent-4-ynoic acid
Formula Weight 213.23
Percent Purity 95%
Chemical Name or Material N-Boc-2-propargyl-D-glycine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.