missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-2-propargyl-L-glycine, 95%, Thermo Scientific™

Product Code. 15468647
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15468647 250 mg 250mg
15458647 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 15468647

Brand: Thermo Scientific Alfa Aesar H52128.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
N-Boc-2-propargyl-L-glycine is used as pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63039-48-5
Molecular Formula C10H15NO4
Molecular Weight (g/mol) 213.233
MDL Number MFCD01320855
InChI Key AMKHAJIFPHJYMH-ZETCQYMHSA-N
Synonym boc-l-propargylglycine, s-2-tert-butoxycarbonyl amino pent-4-ynoic acid, boc-pra-oh, boc-propargyl-gly-oh, n-boc-2-propargyl-l-glycine, 2s-2-tert-butoxycarbonylamino pent-4-ynoic acid, 2s-2-tert-butoxycarbonyl amino pent-4-ynoic acid, 4-pentynoic acid, 2-1,1-dimethylethoxy carbonyl amino-, 2s, boc-propargylglycine, boc-l-propargyl glycine
PubChem CID 2734488
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pent-4-ynoic acid
SMILES CC(C)(C)OC(=O)NC(CC#C)C(=O)O

Specifications

Quantity 250 mg
Solubility Information Soluble in water.
Formula Weight 213.23
Percent Purity 95%
Chemical Name or Material N-Boc-2-propargyl-L-glycine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.