missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-3,4-dichloro-L-phenylalanine, 95%, Thermo Scientific Chemicals

Product Code. 15458547
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15458547 250 mg 250mg
15438547 1 g 1g
15448547 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15458547 Supplier Thermo Scientific Alfa Aesar Supplier No. H52053.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80741-39-5
Molecular Formula C14H16Cl2NO4
Molecular Weight (g/mol) 333.19
MDL Number MFCD00273436
InChI Key UGZIQCCPEDCGGN-NSHDSACASA-M
Synonym boc-3,4-dichloro-l-phenylalanine, boc-phe 3,4-dicl-oh, boc-l-3,4-dichlorophe, s-2-tert-butoxycarbonyl amino-3-3,4-dichlorophenyl propanoic acid, boc-l-3,4-dichlorophenylalanine, 2s-2-tert-butoxycarbonyl amino-3-3,4-dichlorophenyl propanoic acid, 2s-2-tert-butoxycarbonylamino-3-3,4-dichlorophenyl propanoic acid, boc-3,4-dichloro-phe-oh, boc-phe 3,4-cl2-oh, boc-l-3,4-dichloro phenylalanine
PubChem CID 2734501
IUPAC Name (2S)-3-(3,4-dichlorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(Cl)C(Cl)=C1)C([O-])=O

Specifications

Quantity 250 mg
Beilstein 6426101
Formula Weight 334.2
Percent Purity 95%
Chemical Name or Material N-Boc-3,4-dichloro-L-phenylalanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.