missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-3-bromo-2-isoxazoline-5-methylamine, 97%, Thermo Scientific™

Product Code. 11396759
Change view
Click to view available options
Quantity:
500 mg
2.5 g
Unit Size:
2.5g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11396759 2.5 g 2.5g
11386759 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11396759 Supplier Thermo Scientific Alfa Aesar Supplier No. H26274.27
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109770-82-3
Molecular Formula C9H15BrN2O3
Molecular Weight (g/mol) 279.13
MDL Number MFCD08741480
InChI Key AHAHZEKELWWMSJ-UHFFFAOYNA-N
Synonym tert-butyl 3-bromo-4,5-dihydroisoxazol-5-yl methyl carbamate, 3-bromo-4,5-dihydro-isoxazol-5-ylmethyl carbamic acid tert-butyl ester, n-boc-3-bromo-2-isoxazoline-5-methylamine, tert-butyl n-3-bromo-4,5-dihydro-1,2-oxazol-5-yl methyl carbamate, 3-bromo-4,5-dihydro-isoxazol-5-ylmethyl-carbamic acid tert-butyl ester, acmc-20a3tu, tert-butyl 3-bromo-4,5-dihydroisoxazol-5-yl methylcarbamate, 5-n-tert-butoxycarbonyl-aminomethyl-3-bromo-4,5-dihydroisoxazole, tert-butyl 3-bromo-4,5-dihydro-1,2-oxazol-5-yl methyl carbamate
PubChem CID 13637723
IUPAC Name tert-butyl N-[(3-bromo-4,5-dihydro-1,2-oxazol-5-yl)methyl]carbamate
SMILES CC(C)(C)OC(=O)NCC1CC(Br)=NO1

Specifications

Quantity 2.5 g
Formula Weight 279.14
Percent Purity 97%
Chemical Name or Material N-Boc-3-bromo-2-isoxazoline-5-methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.