missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-3-cyclohexyl-D-alanine, 98%

Product Code. 15401568
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15401568 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15401568 Supplier Alfa Aesar™ Supplier No. H55339.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 127095-92-5
Molecular Formula C14H25NO4
Molecular Weight (g/mol) 271.357
MDL Number MFCD00211334
InChI Key MSZQAQJBXGTSHP-LLVKDONJSA-N
Synonym boc-d-cha-oh, r-2-tert-butoxycarbonyl amino-3-cyclohexylpropanoic acid, boc-3-cyclohexyl-d-alanine, boc-d-cha-oh hydrate, 2r-2-tert-butoxycarbonyl amino-3-cyclohexylpropanoic acid, 2r-2-tert-butoxy carbonyl amino-3-cyclohexylpropanoic acid, boc-d-cha, ambotzbaa1314, boc-d-cha-oh*h2o
PubChem CID 2755927
IUPAC Name (2R)-3-cyclohexyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1CCCCC1)C(=O)O

Specifications

Melting Point 64°C to 67°C
Quantity 1 g
Beilstein 4192671
Formula Weight 271.36
Percent Purity 98%
Chemical Name or Material N-Boc-3-cyclohexyl-D-alanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.