missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-3-cyclohexyl-L-alanine, 98%

Product Code. p-7036849
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15471488

Brand: Thermo Scientific Alfa Aesar H55282.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 37736-82-6 | C14H25NO4 | 271.357 g/mol

It is an agrochemical, organic and pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an agrochemical, organic and pharmaceutical intermediates.

Notes
Storage: Keep container tightly closed. Store in a cool, dark and well-ventilated place and away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37736-82-6
Molecular Formula C14H25NO4
Molecular Weight (g/mol) 271.357
MDL Number MFCD00211335
InChI Key MSZQAQJBXGTSHP-NSHDSACASA-N
Synonym boc-cha-oh, boc-cha-oh hydrate, s-2-tert-butoxycarbonyl amino-3-cyclohexylpropanoic acid, boc-beta-cyclohexyl-ala-oh, boc-l-cyclohexylalanine, s-2-tert-butoxycarbonylamino-3-cyclohexyl-propionic acid, n-boc-b-cyclohexylalanine, 2s-2-tert-butoxycarbonyl amino-3-cyclohexylpropanoic acid, boc-cyclohexyl-ala-oh, boc-3-cyclohexyl-l-alanine
PubChem CID 10588189
IUPAC Name (2S)-3-cyclohexyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1CCCCC1)C(=O)O

Specifications

Quantity 250 mg
Beilstein 3060603
Formula Weight 271.36
Percent Purity 98%
Chemical Name or Material N-Boc-3-cyclohexyl-L-alanine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.