missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-4,5-dehydro-L-leucine dicyclohexylamine salt, 97%, Thermo Scientific™

Product Code. 15467645
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15467645 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15467645 Supplier Thermo Scientific Alfa Aesar Supplier No. H27005.MF
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 87720-54-5
Melting Point 148°C to 153°C
Molecular Formula C23H42N2O4
MDL Number MFCD00237524
Quantity 50 mg
UN Number UN3077
Synonym boc-4,5-dehydro-leu-oh dcha, 2s-2-tert-butoxycarbonyl amino-4-methylpent-4-enoic acid; dicha, boc-s-2-methallylglycine dcha, boc-4,5-dehydro-leu-oh . dcha, boc-4,5-dehydro-l-leu-oh . dcha, boc-4,5-dehydro-leu-oh dicyclohexylammonium salt, n-boc-4,5-dehydro-l-leucine dicyclohexylamine salt, boc-4,5-dehydro-leu-oh dicyclohexylammonium salt hplc, dicyclohexylamine s-2-tert-butoxycarbonyl amino-4-methylpent-4-enoate, n-tert-butoxycarbonyl-4-methylidene-l-norvaline-n-cyclohexylcyclohexanamine 1/1
InChI Key ZRIOKRZSXPPREV-FETADZTQNA-N
Optical Rotation +13° (c=1 in Methanol)
SMILES C1CCC(CC1)NC1CCCCC1.CC(=C)C[C@H](NC(=O)OC(C)(C)C)C(O)=O
IUPAC Name N-cyclohexylcyclohexanamine;(2S)-4-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]pent-4-enoic acid
Molecular Weight (g/mol) 410.60
PubChem CID 2755920
Formula Weight 410.59
Percent Purity 97%
Chemical Name or Material N-Boc-4,5-dehydro-L-leucine dicyclohexylamine salt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.