missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-4-oxo-L-proline, 98%

Product Code. 13478648 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 13478648

Brand: Thermo Scientific Acros 449280010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 84348-37-8 | C10H15NO5 | 229.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84348-37-8
Molecular Formula C10H15NO5
Molecular Weight (g/mol) 229.23
MDL Number MFCD01860669
InChI Key CKYGSXRXTIKGAJ-UHFFFAOYNA-N
Synonym n-boc-4-oxo-l-proline, s-1-tert-butoxycarbonyl-4-oxopyrrolidine-2-carboxylic acid, n-t-boc-4-oxo-l-proline, boc-4-oxo-pro-oh, n-tert-boc-4-oxo-l-proline, 2s-1-tert-butoxycarbonyl-4-oxopyrrolidine-2-carboxylic acid, 1-tert-butoxycarbonyl-4-oxoproline, 2s-1-tert-butoxycarbonyl-4-oxo-pyrrolidine-2-carboxylic acid, 2s-1-tert-butoxy carbonyl-4-oxopyrrolidine-2-carboxylic acid, s-4-oxo-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester
PubChem CID 11593804
IUPAC Name (2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]-4-oxopyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CC(=O)CC1C(O)=O

Specifications

Melting Point 155°C to 163°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 229.23
Percent Purity 98%
Chemical Name or Material N-BOC-4-oxo-L-proline
compliance-icons
Product Identifier
  • N-BOC-4-oxo-L-proline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.