missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-4-Piperidinemethanol, 97%

Product Code. 10470894 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10470894 1 g Glass bottle 1g
10585904 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10470894 Supplier Thermo Scientific Acros Supplier No. 397720010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 123855-51-6 | C11H21NO3 | 215.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123855-51-6
Molecular Formula C11H21NO3
Molecular Weight (g/mol) 215.29
MDL Number MFCD02094488
InChI Key CTEDVGRUGMPBHE-UHFFFAOYSA-N
Synonym n-boc-4-piperidinemethanol, tert-butyl 4-hydroxymethyl piperidine-1-carboxylate, 1-boc-4-piperidinemethanol, n-boc-4-hydroxymethylpiperidine, tert-butyl 4-hydroxymethyl-1-piperidinecarboxylate, 1-tert-butoxycarbonyl-4-hydroxymethyl piperidine, 1-n-boc-4-hydroxymethyl-piperidine, n-boc-4-hydroxylmethylene-piperidine, 1-tert-butoxycarbonyl-4-piperidinemethanol, n-tert-butoxycarbonyl-4-piperidinemethanol
PubChem CID 2764081
IUPAC Name tert-butyl 4-(hydroxymethyl)piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(CO)CC1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 78°C to 82°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 215.29
Percent Purity 97%
Physical Form Powder
Chemical Name or Material N-BOC-4-Piperidinemethanol
Show More Show Less
compliance-icons
Product Identifier
  • N-BOC-4-piperidinemethanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.