missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-Amino-(4-N-BOC-piperidinyl)carboxylic acid, 95%, Thermo Scientific™

Product Code. 10520542 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
Packaging:
Glass bottle
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10520542 500 mg Glass bottle 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10520542 Supplier Thermo Scientific Acros Supplier No. 360455000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 189321-65-1
CAS Min % 94.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Molecular Formula C16H27N2O6
MDL Number MFCD02683116
Quantity 500 mg
Synonym 1-tert-butoxycarbonyl-4-tert-butoxycarbonyl amino piperidine-4-carboxylic acid, 1-boc-4-boc-amino piperidine-4-carboxylic acid, n-boc-amino-4-n-boc-piperidinyl carboxylic acid, 1-boc-4-n-boc-amino piperidine-4-carboxylic acid, 4-tert-butoxycarbonylamino-piperidine-1,4-dicarboxylic acid mono-tert-butyl ester, 4-n-boc-amino-4n-boc-piperidinyl-4-carboxylic acid, 1-boc-4-n-boc-amino piperidine-4 carboxylic acid, 1-n-boc-4-n-boc-amino piperidine-4-carboxylic acid, 1-tert-butoxycarbonyl-4-tert-butoxycarbonylamino piperidine-4-carboxylic acid
InChI Key AFEAWYLBSVKOAY-UHFFFAOYSA-M
SMILES CC(C)(C)OC(=O)NC1(CCN(CC1)C(=O)OC(C)(C)C)C([O-])=O
IUPAC Name 1-[(2-methylpropan-2-yl)oxycarbonyl]-4-[(2-methylpropan-2-yl)oxycarbonylamino]piperidine-4-carboxylic acid
Molecular Weight (g/mol) 343.40
PubChem CID 1268225
Formula Weight 344.4
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-BOC-Amino-(4-N-BOC-piperidinyl)carboxylic acid, 95%
Show More Show Less
compliance-icons
Product Identifier
  • N-BOC-Amino-(4-N-BOC-piperidinyl)carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.