missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-beta-alanine, 99%

Product Code. 11470717
Change view
Click to view available options
Quantity:
2.5 g
10 g
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470717 10 g 10g
11460717 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11470717 Supplier Thermo Scientific Alfa Aesar Supplier No. B22522.09
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3303-84-2 | C8H15NO4 | 189.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3303-84-2
Molecular Formula C8H15NO4
Molecular Weight (g/mol) 189.21
MDL Number MFCD00037291
InChI Key WCFJUSRQHZPVKY-UHFFFAOYSA-N
Synonym boc-beta-alanine, boc-beta-ala-oh, n-boc-beta-alanine, boc-b-ala-oh, boc-beta-ala, n-tert-butoxycarbonyl-beta-alanine, 3-tert-butoxycarbonyl amino propanoic acid, n-t-boc-beta-alanine, n-t-butyloxycarbonyl-beta-alanine, 3-tert-butoxycarbonylamino propanoic acid
PubChem CID 76809
IUPAC Name 3-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NCCC(O)=O

Specifications

Melting Point 73°C to 76°C
Linear Formula (CH3)3CO2CNHCH2CH2CO2H
Quantity 10 g
Formula Weight 189.21
Percent Purity 99%
Chemical Name or Material N-Boc-beta-alanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.